ENAH5802E4ED Enamine

methyl 6-amino-2,3-dimethylbenzoate

  • MDL number MFCD20699605
  • Cas Number : 5628-49-9
  • Molecular Weight : 179.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name methyl 6-amino-2,3-dimethylbenzoate
Inchi Code 1S/C10H13NO2/c1-6-4-5-8(11)9(7(6)2)10(12)13-3/h4-5H,11H2,1-3H3
Inchi Key HQXXMRIBSPWVNV-UHFFFAOYSA-N

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-366819

Safety Information

signalword Warning
pcodes P261,P264,P270,P271,P280,P301+P312,P302+P352,P304+P340,P305+P351+P338,P312,P330,P332+P313,P337+P313,P362,P363,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-366819
pictograms GHS07
hstatements H302,H312,H315,H319,H332,H335
Availability details for
Quantity
  • Availability
  • Add to Cart