ENAH5802E4EE Enamine

2-(methylamino)-3-nitrobenzaldehyde

  • MDL number MFCD29120101

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-7440604

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7440604
Availability details for
Quantity
  • Availability
  • Add to Cart