ENAH5802E4EF Enamine

3,3-difluoro-5-methoxy-5-oxopentanoic acid

  • Cas Number : 1779954-64-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7439119

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7439119
Availability details for
Quantity
  • Availability
  • Add to Cart