ENAH5802E4F2 Enamine

methyl 1-(2-aminoethyl)-5-methyl-1H-pyrazole-3-carboxylate dihydrochloride

  • MDL number MFCD32069172
  • Cas Number : 2375268-00-9
  • Molecular Weight : 256.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-7438284

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7438284
Availability details for
Quantity
  • Availability
  • Add to Cart