ENAH5802E4F3 Enamine

ethyl 1-(2-aminoethyl)-1H-pyrazole-3-carboxylate hydrochloride

  • MDL number MFCD31924587
  • Cas Number : 2344679-88-3
  • Molecular Weight : 219.67

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name ethyl 1-(2-aminoethyl)-1H-pyrazole-3-carboxylate hydrochloride
Inchi Code 1S/C8H13N3O2.ClH/c1-2-13-8(12)7-3-5-11(10-7)6-4-9;/h3,5H,2,4,6,9H2,1H3;1H
Inchi Key MPLVPIQDXNTHKB-UHFFFAOYSA-N

Description

Purity 95
Salt Data Cl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-7427173

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7427173
Availability details for
Quantity
  • Availability
  • Add to Cart