ENAH5802E4F4 Enamine

ethyl 2-amino-2-(prop-2-en-1-yl)pent-4-enoate

  • MDL number MFCD20693709
  • Cas Number : 154145-82-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7443546

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7443546
Availability details for
Quantity
  • Availability
  • Add to Cart