ENAH5802E4F8 Enamine

4-(methoxycarbonyl)-1-methyl-1H-pyrazole-3-carboxylic acid

  • MDL number MFCD30377523
  • Cas Number : 1946823-86-4
  • Molecular Weight : 184.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4-(methoxycarbonyl)-1-methyl-1H-pyrazole-3-carboxylic acid
Inchi Code 1S/C7H8N2O4/c1-9-3-4(7(12)13-2)5(8-9)6(10)11/h3H,1-2H3,(H,10,11)
Inchi Key KGSSRKJNMQRGBL-UHFFFAOYSA-N

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-7427021

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7427021
Availability details for
Quantity
  • Availability
  • Add to Cart