ENAH5802E4FA Enamine

methyl 2-[(1r,4r)-4-(aminomethyl)cyclohexyl]acetate hydrochloride

  • MDL number MFCD09951905
  • Cas Number : 2245840-94-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7439737

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7439737
Availability details for
Quantity
  • Availability
  • Add to Cart