ENAH5802E503 Enamine

(2-aminoethyl)[(6-chloropyridin-3-yl)methyl]amine dihydrochloride

  • MDL number MFCD32066296
  • Cas Number : 2361644-57-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7430480

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7430480
Availability details for
Quantity
  • Availability
  • Add to Cart