ENAH5802E504 Enamine

methyl (1r,4r)-4-(aminomethyl)-4-fluorocyclohexane-1-carboxylate hydrochloride

  • MDL number MFCD32068609
  • Cas Number : 2375271-30-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7436314

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7436314
Availability details for
Quantity
  • Availability
  • Add to Cart