ENAH5802E507 Enamine

methyl 6-amino-1-benzofuran-2-carboxylate

  • MDL number MFCD24642656
  • Cas Number : 1167411-36-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-7444537

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7444537
Availability details for
Quantity
  • Availability
  • Add to Cart