ENAH5802E50B Enamine

3-cyano-4-nitrobenzoic acid

  • MDL number MFCD09745179
  • Cas Number : 1350540-03-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-2773878

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-2773878
Availability details for
Quantity
  • Availability
  • Add to Cart