ENAH5802E513 Enamine

methyl 6-amino-1,3-dihydro-2-benzofuran-5-carboxylate

  • MDL number MFCD20724256
  • Cas Number : 872625-35-9
  • Molecular Weight : 193.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name methyl 6-amino-1,3-dihydro-2-benzofuran-5-carboxylate
Inchi Code 1S/C10H11NO3/c1-13-10(12)8-2-6-4-14-5-7(6)3-9(8)11/h2-3H,4-5,11H2,1H3
Inchi Key DERQQAYCYWUIMN-UHFFFAOYSA-N

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-7432541

Safety Information

signalword Warning
pcodes P261,P264,P270,P271,P280,P301+P312,P302+P352,P304+P340,P305+P351+P338,P312,P330,P332+P313,P337+P313,P362,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-7432541
pictograms GHS07
hstatements H302,H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart