ENAH5802E515 Enamine

methyl 3-amino-1,1-dioxo-1lambda6-thiolane-3-carboxylate hydrochloride

  • MDL number MFCD32065971
  • Cas Number : 2361644-71-3
  • Molecular Weight : 229.68

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name methyl 3-aminotetrahydrothiophene-3-carboxylate 1,1-dioxide hydrochloride

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-7431421

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7431421
Availability details for
Quantity
  • Availability
  • Add to Cart