ENAH5802E517 Enamine

ethyl (3S)-3-amino-3-(pyridin-3-yl)propanoate

  • Molecular Weight : 194.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name ethyl (3S)-3-amino-3-(3-pyridinyl)propanoate
Inchi Code 1S/C10H14N2O2/c1-2-14-10(13)6-9(11)8-4-3-5-12-7-8/h3-5,7,9H,2,6,11H2,1H3/t9-/m0/s1
Inchi Key SUOOQSWLTJJSDK-VIFPVBQESA-N

Description

Storage temp. +4
Physical Form liquid
Shipping Temperature Ice Pack
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-2174549

Safety Information

signalword Danger
pcodes P261,P264,P270,P271,P280,P301+P312,P302+P352,P304+P340,P305+P351+P338,P310,P312,P330,P332+P313,P362,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-2174549
pictograms GHS05,GHS07
hstatements H302,H315,H318,H335
Availability details for
Quantity
  • Availability
  • Add to Cart