ENAH5802E518 Enamine

2-formyl-6H-thieno[2,3-b]pyrrole-5-carboxylic acid

  • MDL number MFCD20642455
  • Cas Number : 58963-45-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-702619

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-702619
Availability details for
Quantity
  • Availability
  • Add to Cart