ENAH5802E51A Enamine

methyl 2-(aminomethyl)-4H,5H,6H-pyrrolo[1,2-b]pyrazole-3-carboxylate hydrochloride

  • MDL number MFCD32198439
  • Cas Number : 2375269-88-6
  • Molecular Weight : 231.68

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name methyl 2-(aminomethyl)-5,6-dihydro-4H-pyrrolo[1,2-b]pyrazole-3-carboxylate hydrochloride

Description

Purity 95
Salt Data Cl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-7439955

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7439955
Availability details for
Quantity
  • Availability
  • Add to Cart