ENAH5802E51C Enamine

5-chloroindolizine-2-carboxylic acid

  • MDL number MFCD32066896
  • Molecular Weight : 195.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 5-chloroindolizine-2-carboxylic acid
Inchi Code 1S/C9H6ClNO2/c10-8-3-1-2-7-4-6(9(12)13)5-11(7)8/h1-5H,(H,12,13)
Inchi Key GSTLMKYKSLPFLF-UHFFFAOYSA-N

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-1662996

Safety Information

signalword Warning
pcodes P261,P264,P270,P271,P280,P301+P312,P302+P352,P304+P340,P305+P351+P338,P312,P330,P332+P313,P337+P313,P362,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-1662996
pictograms GHS07
hstatements H302,H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart