ENAH5802E521 Enamine

methyl 2-amino-3-(1,5-dimethyl-1H-pyrazol-4-yl)propanoate dihydrochloride

  • MDL number MFCD32198447
  • Cas Number : 2375260-22-1
  • Molecular Weight : 270.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-7443307

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7443307
Availability details for
Quantity
  • Availability
  • Add to Cart