ENAH5802E525 Enamine

methyl 1-(azetidin-3-yl)-5-oxopyrrolidine-3-carboxylate hydrochloride

  • MDL number MFCD32066244
  • Cas Number : 2361634-21-9
  • Molecular Weight : 234.68

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name methyl 1-(azetidin-3-yl)-5-oxopyrrolidine-3-carboxylate hydrochloride
Inchi Code 1S/C9H14N2O3.ClH/c1-14-9(13)6-2-8(12)11(5-6)7-3-10-4-7;/h6-7,10H,2-5H2,1H3;1H
Inchi Key UMFJWZYYTGNCKL-UHFFFAOYSA-N

Description

Purity 95
Salt Data Cl
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-7430283

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7430283
Availability details for
Quantity
  • Availability
  • Add to Cart