ENAH5802E526 Enamine

tert-butyl 4-amino-2-azabicyclo[3.1.0]hexane-2-carboxylate

  • Cas Number : 2344685-22-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7432098

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7432098
Availability details for
Quantity
  • Availability
  • Add to Cart