ENAH5802E529 Enamine

tert-butyl 4-amino-2-azabicyclo[2.1.1]hexane-2-carboxylate

  • MDL number MFCD32066114
  • Cas Number : 2361636-63-5
  • Molecular Weight : 198.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name tert-butyl 4-amino-2-azabicyclo[2.1.1]hexane-2-carboxylate
Inchi Code 1S/C10H18N2O2/c1-9(2,3)14-8(13)12-6-10(11)4-7(12)5-10/h7H,4-6,11H2,1-3H3
Inchi Key FQLACRHRFMGHJC-UHFFFAOYSA-N

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-7434257

Safety Information

signalword Warning
pcodes P261,P264,P270,P271,P280,P301+P312,P302+P352,P304+P340,P305+P351+P338,P312,P330,P332+P313,P337+P313,P362,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-7434257
pictograms GHS07
hstatements H302,H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart