ENAH5802E531 Enamine

4,4-dimethyl piperidine-4,4-dicarboxylate hydrochloride

  • MDL number MFCD32066105
  • Cas Number : 2361643-41-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7434168

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7434168
Availability details for
Quantity
  • Availability
  • Add to Cart