ENAH5802E537 Enamine

2-chloro-6-(hydroxymethyl)pyridine-4-carboxylic acid

  • MDL number MFCD20642054
  • Cas Number : 900799-89-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-7442518

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7442518
Availability details for
Quantity
  • Availability
  • Add to Cart