ENAH5802E53A Enamine

methyl 2-amino-6,6-difluorospiro[3.3]heptane-2-carboxylate hydrochloride

  • MDL number MFCD32066107
  • Cas Number : 2361644-03-1
  • Molecular Weight : 241.67

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name methyl 2-amino-6,6-difluorospiro[3.3]heptane-2-carboxylate hydrochloride
Inchi Code 1S/C9H13F2NO2.ClH/c1-14-6(13)8(12)2-7(3-8)4-9(10,11)5-7;/h2-5,12H2,1H3;1H
Inchi Key DCDBKFWVTZFEBK-UHFFFAOYSA-N

Description

Purity 95
Salt Data Cl
Storage temp. +4
Physical Form powder
Shipping Temperature Ice Pack
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-7434178

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7434178
Availability details for
Quantity
  • Availability
  • Add to Cart