ENAH5802E53D Enamine

methyl 2,3,4,5-tetrahydro-1H-1-benzazepine-4-carboxylate

  • MDL number MFCD32065941
  • Cas Number : 2092720-37-9
  • Molecular Weight : 205.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name methyl 2,3,4,5-tetrahydro-1H-benzo[b]azepine-4-carboxylate
Inchi Code 1S/C12H15NO2/c1-15-12(14)10-6-7-13-11-5-3-2-4-9(11)8-10/h2-5,10,13H,6-8H2,1H3
Inchi Key LHUHOYMOLWBMSW-UHFFFAOYSA-N

Description

Purity 95
Storage temp. +4
Physical Form oil
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-7433795

Safety Information

signalword Warning
pcodes P261,P264,P270,P271,P280,P301+P312,P302+P352,P304+P340,P305+P351+P338,P312,P330,P332+P313,P337+P313,P362,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-7433795
pictograms GHS07
hstatements H302,H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart