ENAH5802E541 Enamine

methyl (2S,4R)-4-methanesulfonylpyrrolidine-2-carboxylate hydrochloride

  • MDL number MFCD32198034
  • Cas Number : 2375248-23-8
  • Molecular Weight : 243.71

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-7443059

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7443059
Availability details for
Quantity
  • Availability
  • Add to Cart