ENAH5802E542 Enamine

ethyl 2-amino-3-(3-methylphenyl)propanoate

  • MDL number MFCD05228070

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-7442969

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7442969
Availability details for
Quantity
  • Availability
  • Add to Cart