ENAH5802E545 Enamine

2-(methoxycarbonyl)-3,6-dimethylbenzoic acid

  • MDL number MFCD24487375
  • Cas Number : 1015-54-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-5336062

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-5336062
Availability details for
Quantity
  • Availability
  • Add to Cart