ENAH5802E546 Enamine

methyl 5-aminothieno[2,3-d]pyrimidine-6-carboxylate

  • MDL number MFCD32197836
  • Cas Number : 2091691-82-4
  • Molecular Weight : 209.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name methyl 5-aminothieno[2,3-d]pyrimidine-6-carboxylate
Inchi Code 1S/C8H7N3O2S/c1-13-8(12)6-5(9)4-2-10-3-11-7(4)14-6/h2-3H,9H2,1H3
Inchi Key WMSOTJQOBVVWJM-UHFFFAOYSA-N

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-7441034

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7441034
Availability details for
Quantity
  • Availability
  • Add to Cart