ENAH5802E547 Enamine

ethyl 5-methyl-4H,5H,6H,7H-pyrazolo[1,5-a]pyrimidine-3-carboxylate

  • MDL number MFCD29487019
  • Cas Number : 1699153-63-3
  • Molecular Weight : 209.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name ethyl 5-methyl-4,5,6,7-tetrahydropyrazolo[1,5-a]pyrimidine-3-carboxylate
Inchi Code 1S/C10H15N3O2/c1-3-15-10(14)8-6-11-13-5-4-7(2)12-9(8)13/h6-7,12H,3-5H2,1-2H3
Inchi Key LYUNDNQGNKVLCT-UHFFFAOYSA-N

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-367110

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-367110
Availability details for
Quantity
  • Availability
  • Add to Cart