ENAH5802E550 Enamine

2-[4-(ethoxycarbonyl)-5-imino-2,5-dihydro-1H-pyrazol-1-yl]acetic acid

  • MDL number MFCD26143905
  • Cas Number : 1355694-40-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-70823

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-70823
Availability details for
Quantity
  • Availability
  • Add to Cart