ENAH5802E552 Enamine

methyl 3-oxa-9-azaspiro[5.5]undecane-7-carboxylate hydrochloride

  • MDL number MFCD32068922
  • Cas Number : 2361646-52-6
  • Molecular Weight : 249.74

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-7434614

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7434614
Availability details for
Quantity
  • Availability
  • Add to Cart