ENAH5802E556 Enamine

2-bromo-4-cyclopropyl-1,3-oxazole

  • MDL number MFCD28151775
  • Cas Number : 1602201-26-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-310709

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-310709
Availability details for
Quantity
  • Availability
  • Add to Cart