ENAH5802E564 Enamine

ethyl 2-(2-aminoacetamido)-4-methylpentanoate hydrochloride

  • MDL number MFCD32065985
  • Cas Number : 2087-46-9
  • Molecular Weight : 252.74

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name ethyl glycylleucinate hydrochloride
Inchi Code 1S/C10H20N2O3.ClH/c1-4-15-10(14)8(5-7(2)3)12-9(13)6-11;/h7-8H,4-6,11H2,1-3H3,(H,12,13);1H
Inchi Key SBIYAKXNZXDPAJ-UHFFFAOYSA-N

Description

Purity 95
Salt Data Cl
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-7433798

Safety Information

signalword Warning
pcodes P261,P264,P271,P280,P302+P352,P304+P340,P305+P351+P338,P312,P332+P313,P337+P313,P362,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-7433798
pictograms GHS07
hstatements H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart