ENAH5802E567 Enamine

8-nitroquinoline-2-carboxylic acid

  • MDL number MFCD06742777
  • Cas Number : 15733-85-4
  • Molecular Weight : 218.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-729613

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-729613
Availability details for
Quantity
  • Availability
  • Add to Cart