ENAH5802E56A Enamine

methyl 3,4-dihydrospiro[1,4-benzoxazine-2,1'-cyclopropane]-8-carboxylate

  • MDL number MFCD32197665
  • Cas Number : 2375270-22-5
  • Molecular Weight : 219.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-7436241

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7436241
Availability details for
Quantity
  • Availability
  • Add to Cart