ENAH5802E573 Enamine

methyl 2-[4-(2-aminopropyl)phenoxy]acetate hydrochloride

  • MDL number MFCD32066168
  • Cas Number : 114962-90-2
  • Molecular Weight : 259.73

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 123-125
Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-7434346

Safety Information

signalword Warning
pcodes P261,P264,P271,P280,P302+P352,P304+P340,P305+P351+P338,P312,P332+P313,P337+P313,P362,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-7434346
pictograms GHS07
hstatements H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart