ENAH5802E596 Enamine

tert-butyl 3-(aminomethyl)-3-(methoxymethyl)azetidine-1-carboxylate

  • MDL number MFCD28631503
  • Cas Number : 1781613-26-0
  • Molecular Weight : 230.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Ice Pack
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-6929874

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6929874
Availability details for
Quantity
  • Availability
  • Add to Cart