ENAH5802E5AB Enamine

ethyl 3-imino-2-(piperidin-4-yl)-2,3-dihydro-1H-pyrazole-4-carboxylate dihydrochloride

  • MDL number MFCD28155356
  • Cas Number : 1355773-02-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Product Link https://www.enaminestore.com/catalog/EN300-78422

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-78422
Availability details for
Quantity
  • Availability
  • Add to Cart