ENAH5802E5C4 Enamine

tert-butyl 5-(aminomethyl)-3,3-dimethylpiperidine-1-carboxylate

  • MDL number MFCD32069246
  • Cas Number : 1785324-09-5
  • Molecular Weight : 242.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-7167268

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7167268
Availability details for
Quantity
  • Availability
  • Add to Cart