ENAH5802E66B Enamine

2-{[(tert-butoxy)carbonyl]amino}spiro[3.5]nonane-2-carboxylic acid

  • MDL number MFCD32197948
  • Cas Number : 2375261-36-0
  • Molecular Weight : 283.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-7444539

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7444539
Availability details for
Quantity
  • Availability
  • Add to Cart