ENAH5802E78D Enamine

2H-1,3-dithiole-4-carboxylic acid

  • MDL number MFCD18805944
  • Cas Number : 58758-32-0
  • Molecular Weight : 148.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -10
Physical Form powder
Shipping Temperature Ice Pack
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-7352416

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7352416
Availability details for
Quantity
  • Availability
  • Add to Cart