ENAH5802E7B2 Enamine

(1s,4s)-4-fluoro-4-methylcyclohexane-1-carboxylic acid

  • Cas Number : 1027708-67-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7434879

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7434879
Availability details for
Quantity
  • Availability
  • Add to Cart