ENAH5802E7BA Enamine

sodium [1,2]oxazolo[5,4-c]pyridine-3-carboxylate

  • Cas Number : 2344680-41-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data Na+
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6745638

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6745638
Availability details for
Quantity
  • Availability
  • Add to Cart