ENAH5802E7BE Enamine

5-cyclopropylpyridine-3-carboxylic acid

  • MDL number MFCD06802408
  • Cas Number : 893738-23-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1221121

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1221121
Availability details for
Quantity
  • Availability
  • Add to Cart