ENAH5802E7C2 Enamine

1-methyl-1H,4H-pyrrolo[2,3-d]imidazole-5-carboxylic acid

  • MDL number MFCD19688273
  • Cas Number : 1368352-84-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-120964

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-120964
Availability details for
Quantity
  • Availability
  • Add to Cart