ENAH5802E7C3 Enamine

5-(propan-2-yl)pyridine-3-carboxylic acid hydrochloride

  • Cas Number : 2375269-80-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7441793

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7441793
Availability details for
Quantity
  • Availability
  • Add to Cart