ENAH5802E7CD Enamine

3-{[(tert-butoxy)carbonyl](methyl)amino}-4,4-difluorobutanoic acid

  • MDL number MFCD32068804
  • Cas Number : 1781342-40-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1709285

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1709285
Availability details for
Quantity
  • Availability
  • Add to Cart