ENAH5802E7D0 Enamine

4H,5H,6H,7H-[1,2,3]triazolo[1,5-a]pyridine-5-carboxylic acid

  • MDL number MFCD27986488
  • Cas Number : 1538333-18-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-748882

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-748882
Availability details for
Quantity
  • Availability
  • Add to Cart